Merck
  • No relevant cardiac, pharmacokinetic or safety interactions between roflumilast and inhaled formoterol in healthy subjects: an open-label, randomised, actively controlled study.
No relevant cardiac, pharmacokinetic or safety interactions between roflumilast and inhaled formoterol in healthy subjects: an open-label, randomised, actively controlled study.
BMC clinical pharmacology (2011-06-03)
Christian de Mey, Nassr Nassr, Gezim Lahu
ABSTRACT
Roflumilast is an oral, selective phosphodiesterase 4 inhibitor with anti-inflammatory effects in chronic obstructive pulmonary disease (COPD). The addition of roflumilast to long-acting bronchodilators improves lung function in patients with moderate-to-severe COPD. The present study investigated drug-drug interaction effects between inhaled formoterol and oral roflumilast. This was a single-centre (investigational clinic), open, randomised, multiple-dose, parallel-group study. In Regimen A, healthy men were treated with roflumilast (500 μg tablet once daily; Day 2-18) and concomitant formoterol (24 μg twice daily; Day 12-18). In Regimen B, healthy men were treated with formoterol (24 μg twice daily; Day 2-18) and concomitant roflumilast (500 μg once daily; Day 9-18). Steady-state plasma pharmacokinetics of roflumilast, roflumilast N-oxide and/or formoterol (Cmax and AUC0-τ) as well as pharmacodynamics - blood pressure, transthoracic impedance cardiography (ZCG), 12-lead digital electrocardiography, peripheral blood eosinophils, and serum glucose and potassium concentrations - were evaluated through Day 1 (baseline), Day 8 (Regimen B: formoterol alone) or Day 11 (Regimen A: roflumilast alone), and Day 18 (Regimen A and B: roflumilast plus formoterol). Blood and urine samples were taken for safety assessment at screening, pharmacokinetic profiling days and Day 19. Adverse events were monitored throughout the study. Of the 27 subjects enrolled, 24 were evaluable (12 in each regimen). No relevant pharmacokinetic interactions occurred. Neither roflumilast nor formoterol were associated with significant changes in cardiovascular parameters as measured by ZCG, and these parameters were not affected during concomitant administration. Formoterol was associated with a slight increase in heart rate and a corresponding shortening of the QT interval, without changes in the heart rate-corrected QTc interval. There were small effects on the other pharmacodynamic assessments when roflumilast and formoterol were administered individually, but no interactions or safety concerns were seen after concomitant administration. No severe or serious adverse events were reported, and no adverse events led to premature study discontinuation. No clinically relevant pharmacokinetic or pharmacodynamic interactions were found when oral roflumilast was administered concomitantly with inhaled formoterol, including no effect on cardiac repolarisation. Roflumilast was well tolerated.
Sigma-Aldrich
Roflumilast, ≥98% (HPLC)