Merck
  • Phosphorylation of DARPP-32 by Cdk5 modulates dopamine signalling in neurons.
Phosphorylation of DARPP-32 by Cdk5 modulates dopamine signalling in neurons.
Nature (1999-12-22)
J A Bibb, G L Snyder, A Nishi, Z Yan, L Meijer, A A Fienberg, L H Tsai, Y T Kwon, J A Girault, A J Czernik, R L Huganir, H C Hemmings, A C Nairn, P Greengard
ABSTRACT
The physiological state of the cell is controlled by signal transduction mechanisms which regulate the balance between protein kinase and protein phosphatase activities. Here we report that a single protein can, depending on which particular amino-acid residue is phosphorylated, function either as a kinase or phosphatase inhibitor. DARPP-32 (dopamine and cyclic AMP-regulated phospho-protein, relative molecular mass 32,000) is converted into an inhibitor of protein phosphatase 1 when it is phosphorylated by protein kinase A (PKA) at threonine 34. We find that DARPP-32 is converted into an inhibitor of PKA when phosphorylated at threonine 75 by cyclin-dependent kinase 5 (Cdk5). Cdk5 phosphorylates DARPP-32 in vitro and in intact brain cells. Phospho-Thr 75 DARPP-32 inhibits PKA in vitro by a competitive mechanism. Decreasing phospho-Thr 75 DARPP-32 in striatal slices, either by a Cdk5-specific inhibitor or by using genetically altered mice, results in increased dopamine-induced phosphorylation of PKA substrates and augmented peak voltage-gated calcium currents. Thus DARPP-32 is a bifunctional signal transduction molecule which, by distinct mechanisms, controls a serine/threonine kinase and a serine/threonine phosphatase.