Merck
  • An estrogen receptor dependent mechanism of Oroxylin A in the repression of inflammatory response.
An estrogen receptor dependent mechanism of Oroxylin A in the repression of inflammatory response.
PloS one (2013-08-08)
Hong Wang, Ying Guo, Xin Zhao, Huiying Li, Guanwei Fan, Haoping Mao, Lin Miao, Xiumei Gao
ABSTRACT
Oroxylin A, a natural flavonoid, is one of the main bioactive compounds that underlie the anti-inflammatory effect of the medicinal herb Scutellaria baicalensis Georgi widely used in southeastern Asia; however, the molecular mechanisms for the therapeutic benefits remain largely unclear. In this study, we found that Oroxylin A induces estrogen-responsive gene expression and promoter activity. In macrophages, Oroxylin A treatment significantly attenuates lipopolysaccharide (LPS)-induced but not basal inflammatory response, including nitric oxide (NO) production and the expression of inflammatory mediators (i.e., iNOS and COX-2) and cytokines (i.e., TNF-α, IL-1β, and IL-6), in an estrogen receptor (ER)-dependent manner. Oroxylin A treatment also dramatically decreases LPS-induced secretion of pro-inflammatory cytokines. Furthermore, the downregulation of all these inflammatory parameters by Oroxylin A was abolished when cells were pretreated with specific ER antagonist. Thus, Oroxylin A is a novel phytoestrogen and exhibits anti-inflammatory effects that are mediated by ER activity.