Merck
  • Matrix metalloproteinase inhibitors containing a (carboxyalkyl)amino zinc ligand: modification of the P1 and P2' residues.
Matrix metalloproteinase inhibitors containing a (carboxyalkyl)amino zinc ligand: modification of the P1 and P2' residues.
Journal of medicinal chemistry (1994-03-04)
F K Brown, P J Brown, D M Bickett, C L Chambers, H G Davies, D N Deaton, D Drewry, M Foley, A B McElroy, M Gregson
ABSTRACT
Systematic modification of the presumed P1 side chain in a series of (carboxyalkyl)amino-based inhibitors of matrix metalloproteinases enabled identification of the 2-(1,3-dihydro-1,3-dioxo-2H-benz[f]isoindol-2-yl)ethyl group as a preferred substituent imparting potent inhibition of the enzymes collagenase and gelatinase. It was subsequently found that the P2'-P3' residues in this series could be replaced by small non-peptide residues, while maintaining inhibitory potency. The imide group in this series of compounds can undergo autocatalytic hydrolysis under neutral conditions.