Merck
  • Autophagy regulates Notch degradation and modulates stem cell development and neurogenesis.
Autophagy regulates Notch degradation and modulates stem cell development and neurogenesis.
Nature communications (2016-02-04)
Xiaoting Wu, Angeleen Fleming, Thomas Ricketts, Mariana Pavel, Herbert Virgin, Fiona M Menzies, David C Rubinsztein
ABSTRACT
Autophagy is a conserved, intracellular, lysosomal degradation pathway. While mechanistic aspects of this pathway are increasingly well defined, it remains unclear how autophagy modulation impacts normal physiology. It is, however, becoming clear that autophagy may play a key role in regulating developmental pathways. Here we describe for the first time how autophagy impacts stem cell differentiation by degrading Notch1. We define a novel route whereby this plasma membrane-resident receptor is degraded by autophagy, via uptake into ATG16L1-positive autophagosome-precursor vesicles. We extend our findings using a physiologically relevant mouse model with a hypomorphic mutation in Atg16L1, a crucial autophagy gene, which shows developmental retention of early-stage cells in various tissues where the differentiation of stem cells is retarded and thus reveal how modest changes in autophagy can impact stem cell fate. This may have relevance for diverse disease conditions, like Alzheimer's Disease or Crohn's Disease, associated with altered autophagy.
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
ANTI-GREMLIN (C-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution