Merck
  • Intermittent Stretching and Osteogenic Differentiation of Bone Marrow Derived Mesenchymal Stem Cells via the p38MAPK-Osterix Signaling Pathway.
Intermittent Stretching and Osteogenic Differentiation of Bone Marrow Derived Mesenchymal Stem Cells via the p38MAPK-Osterix Signaling Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-06-27)
Wen-lin Xiao, Dai-zun Zhang, Cun-hui Fan, Bao-jun Yu
ABSTRACT
The relationship between the p38MAPK signaling pathway and osterix in osteogenic differentiation of BMMSCs subjected to intermittent stretching was investigated. BMMSCs derived from C57BL/6J mice were divided into the following groups: 1) control, 2) stretch, and 3) SB203580+stretch (SB203580 is a p38MAPK signal pathway inhibitor). BMMSCs were exposed to an intermittent mechanical strain of 0.8% (8000μ strain) at 0.5 Hz, twice a day for 30 min each application. BMMSCs were harvested on days 1, 3, and 5 post-treatment. The expression of ALP, COL I, OCN, and osterix mRNA was assessed utilizing RT-PCR while the expression of P-p38MAPK and osterix protein was assessed by Western blot analysis. The osterix gene in mouse BMMSCs was knocked down using RNAi technology and its protein expression was also assessed by Western blot. RT-PCR was used to detect ALP, COL I, and OCN mRNA expression. Intermittent stretching was found to promote expression of ALP, COL I, OCN, and osterix mRNA. Silencing the osterix gene was found to reduce levels of ALP, COL I, and OCN mRNA. Western blot analysis demonstrated that the levels of osterix and P-p38MAPK proteins in the stretch group were significantly higher than in the control group (P<0.05). There was less expression of ALP, COL I, OCN, and osterix mRNA in the SB203580+stretch group than in the control and stretch groups. Data demonstrate that intermittent stretching promotes osteogenic differentiation of BMMSCs, and the p38MAPK-osterix pathway has an important role in the control of osteogenesis-related gene expression.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sp7
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Alizarin, Dye content 97 %