Merck
  • Analysis of celiac disease autoreactive gut plasma cells and their corresponding memory compartment in peripheral blood using high-throughput sequencing.
Analysis of celiac disease autoreactive gut plasma cells and their corresponding memory compartment in peripheral blood using high-throughput sequencing.
Journal of immunology (Baltimore, Md. : 1950) (2015-05-15)
Omri Snir, Luka Mesin, Moriah Gidoni, Knut E A Lundin, Gur Yaari, Ludvig M Sollid
ABSTRACT
Autoreactive IgA plasma cells (PCs) specific for the enzyme transglutaminase 2 (TG2) are abundant in the small intestine of patients with active celiac disease (CD), and their number drops in patients treated by dietary gluten elimination. Little is known about their characteristics and their role in the disease. In this study, using high-throughput sequencing of the IgH V region (IGHV) genes, we have studied features of TG2-specific PCs and their related B cell clones in peripheral blood. We found that TG2-specific PCs from both untreated and treated patients have acquired lower number of somatic hypermutation and used focused IGHV repertoire with overrepresentation of the IGHV3-48, IGHV4-59, IGHV5-10-1, and IGHV5-51 gene segments. Furthermore, these PCs were clonally expanded and showed signs of affinity maturation. Lineage trees demonstrated shared clones between gut PCs and blood memory B cells, primarily IgAs. Some trees also involved IgG cells, suggesting that anti-TG2 IgA and IgG responses are related. Similarly to TG2-specific PCs, clonally related memory IgA B cells of blood showed lower mutation rates with biased usage of IGHV3-48 and IGHV5-51. Such memory cells were rare in peripheral blood, yet detectable in most patients assessed by production of anti-TG2 Abs in vitro following stimulation of cells from patients who had been on a long-term gluten-free diet. Thus, the Ab response to TG2 in CD, while maintaining its IGHV gene usage, is dynamically regulated in response to gluten exposure with a low degree of maintenance at both PC and memory B cell levels in patients in remission.
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type H, ≥1.0 FALGPA units/mg solid
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Phosphatase substrate, Suitable for manufacturing of diagnostic kits and reagents
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder