Merck
  • Identification of DRG-1 As a Melanoma-Associated Antigen Recognized by CD4+ Th1 Cells.
Identification of DRG-1 As a Melanoma-Associated Antigen Recognized by CD4+ Th1 Cells.
PloS one (2015-05-21)
Yukiko Kiniwa, Jiang Li, Mingjun Wang, Chuang Sun, Jeffrey E Lee, Rong-Fu Wang, Helen Y Wang
ABSTRACT
Immunotherapy has emerged as a promising strategy for the treatment of metastatic melanoma. Clinical studies have demonstrated the feasibility of cancer immunotherapy using tumor antigens recognized by CD8(+) T cells. However, the overall immune responses induced by these antigens are too weak and transient to induce tumor regression in the majority of patients who received immunization. A growing body of evidence suggests that CD4(+) T helper (Th) cells play an important role in antitumor immunity. Therefore, the identification of MHC class II-restricted tumor antigens capable of stimulating CD4(+) T cells may provide opportunities for developing effective cancer vaccines. To this end, we describe the identification of developmentally regulated GTP-binding protein 1 (DRG-1) as a melanoma-associated antigen recognized by HLA-DR11-restricted CD4(+) Th1 cells. Epitope mapping analysis showed that the DRG1248-268 epitope of DRG-1 was required for T cell recognition. Reverse transcription-polymerase chain reaction revealed that DRG-1 was highly expressed in melanoma cell lines but not in normal tissues. DRG-1 knockdown by lentiviral-based shRNA suppressed melanoma cell proliferation and soft agar colony formation. Taken together, these data suggest that DRG-1 plays an important role in melanoma cell growth and transformation, indicating that DRG1 may represent a novel target for CD4(+) T cell-mediated immunotherapy in melanoma.
Sigma-Aldrich
Thiazole, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Thiazole, ≥99%
Pricing and availability is not currently available.