Merck
  • Parmodulins inhibit thrombus formation without inducing endothelial injury caused by vorapaxar.
Parmodulins inhibit thrombus formation without inducing endothelial injury caused by vorapaxar.
Blood (2015-01-15)
Omozuanvbo Aisiku, Christian G Peters, Karen De Ceunynck, Chandra C Ghosh, James R Dilks, Susanna F Fustolo-Gunnink, Mingdong Huang, Chris Dockendorff, Samir M Parikh, Robert Flaumenhaft
ABSTRACT
Protease-activated receptor-1 (PAR1) couples the coagulation cascade to platelet activation during myocardial infarction and to endothelial inflammation during sepsis. This receptor demonstrates marked signaling bias. Its activation by thrombin stimulates prothrombotic and proinflammatory signaling, whereas its activation by activated protein C (APC) stimulates cytoprotective and antiinflammatory signaling. A challenge in developing PAR1-targeted therapies is to inhibit detrimental signaling while sparing beneficial pathways. We now characterize a novel class of structurally unrelated small-molecule PAR1 antagonists, termed parmodulins, and compare the activity of these compounds to previously characterized compounds that act at the PAR1 ligand-binding site. We find that parmodulins target the cytoplasmic face of PAR1 without modifying the ligand-binding site, blocking signaling through Gαq but not Gα13 in vitro and thrombus formation in vivo. In endothelium, parmodulins inhibit prothrombotic and proinflammatory signaling without blocking APC-mediated pathways or inducing endothelial injury. In contrast, orthosteric PAR1 antagonists such as vorapaxar inhibit all signaling downstream of PAR1. Furthermore, exposure of endothelial cells to nanomolar concentrations of vorapaxar induces endothelial cell barrier dysfunction and apoptosis. These studies demonstrate how functionally selective antagonism can be achieved by targeting the cytoplasmic face of a G-protein-coupled receptor to selectively block pathologic signaling while preserving cytoprotective pathways.
Sigma-Aldrich
MISSION® esiRNA, targeting human F2R
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Gpr172b
Sigma-Aldrich
MISSION® esiRNA, targeting human NR1I2