Merck
  • Lowered nudix type 5 (NUDT5) expression leads to cell cycle retardation in HeLa cells.
Lowered nudix type 5 (NUDT5) expression leads to cell cycle retardation in HeLa cells.
Molecular and cellular biochemistry (2011-12-28)
Li-Qun Zhang, Da-Peng Dai, Wei Gan, Yasumitsu Takagi, Hiroshi Hayakawa, Mutsuo Sekiguchi, Jian-Ping Cai
ABSTRACT
The molecule 8-oxo-7,8-dihydroguanine (8-oxoGua), an oxidized form of guanine, can pair with adenine or cytosine during nucleic acid synthesis. Moreover, RNA containing 8-oxoGua causes translational errors, thus leading to the production of abnormal proteins. Human NUDT5, a MutT-related protein, catalyzes the hydrolysis of 8-oxoGDP to 8-oxoGMP, thereby preventing misincorporation of 8-oxoGua into RNA. To investigate the biological roles of NUDT5 in mammalian cells, we established cell lines with decreased level of NUDT5 expression. In NUDT5 inhibited cells, the RNA oxidation was not significantly higher than that of normal cells. However, the cell cycle G1 phase was significantly delayed, and cell numbers in both S and G2/M phases were reduced, indicating that cell proliferation was hampered by NUDT5 suppression. Key proteins for preventing the G1-S transition, including p53, p16, and Rb were increased, while the Rb phosphorylation was decreased. These results suggested that the NUDT5 protein may play significant roles in regulating the G1-S transition in mammalian cells.