Merck
  • SMAD4 suppresses AURKA-induced metastatic phenotypes via degradation of AURKA in a TGFβ-independent manner.
SMAD4 suppresses AURKA-induced metastatic phenotypes via degradation of AURKA in a TGFβ-independent manner.
Molecular cancer research : MCR (2014-07-26)
Lina Jia, Hun Seok Lee, Chun Fu Wu, Juthika Kundu, Sang Gyu Park, Ryong Nam Kim, Li-Hui Wang, Özgür Cem Erkin, Jong-Sun Choi, Seoung Wan Chae, Ho Bin Yang, Yoon-La Choi, Young Kee Shin
ABSTRACT
SMAD4 has been suggested to inhibit the activity of the WNT/β-catenin signaling pathway in cancer. However, the mechanism by which SMAD4 antagonizes WNT/β-catenin signaling in cancer remains largely unknown. Aurora A kinase (AURKA), which is frequently overexpressed in cancer, increases the transcriptional activity of β-catenin/T-cell factor (TCF) complex by stabilizing β-catenin through the inhibition of GSK-3β. Here, SMAD4 modulated AURKA in a TGFβ-independent manner. Overexpression of SMAD4 significantly suppressed AURKA function, including colony formation, migration, and invasion of cell lines. In addition, SMAD4 bound to AURKA induced degradation of AURKA by the proteasome. A luciferase activity assay revealed that the transcriptional activity of the β-catenin/TCF complex was elevated by AURKA, but decreased by SMAD4 overexpression. Moreover, target gene analysis showed that SMAD4 abrogated the AURKA-mediated increase of β-catenin target genes. However, this inhibitory effect of SMAD4 was abolished by overexpression of AURKA or silencing of AURKA in SMAD4-overexpressed cells. Meanwhile, the SMAD4-mediated repression of AURKA and β-catenin was independent of TGFβ signaling because blockage of TGFβR1 or restoration of TGFβ signaling did not prevent suppression of AURKA and β-catenin signaling by SMAD4. These results indicate that the tumor-suppressive function of SMAD4 is mediated by downregulation of β-catenin transcriptional activity via AURKA degradation in a TGFβ-independent manner. SMAD4 interacts with AURKA and antagonizes its tumor-promoting potential, thus demonstrating a novel mechanism of tumor suppression.