Merck
CCN3 (NOV) is a novel angiogenic regulator of the CCN protein family.
The Journal of biological chemistry (2003-04-16)
Cristiane G Lin, Shr-Jeng Leu, Ningyu Chen, Christopher M Tebeau, Shao-Xia Lin, Cho-Yau Yeung, Lester F Lau
ABSTRACT
CCN3 (NOV) is a matricellular protein of the CCN family, which also includes CCN1 (CYR61), CCN2 (CTGF), CCN4 (WISP-1), CCN5 (WISP-2), and CCN6 (WISP-3). During development, CCN3 is expressed widely in derivatives of all three germ layers, and high levels of expression are observed in smooth muscle cells of the arterial vessel wall. Altered expression of CCN3 has been observed in a variety of tumors, including hepatocellular carcinomas, Wilm's tumors, Ewing's sarcomas, gliomas, rhabdomyosarcomas, and adrenocortical carcinomas. To understand its biological functions, we have investigated the activities of purified recombinant CCN3. We show that in endothelial cells, CCN3 supports cell adhesion, induces directed cell migration (chemotaxis), and promotes cell survival. Mechanistically, CCN3 supports human umbilical vein endothelial cell adhesion through multiple cell surface receptors, including integrins alphavbeta3, alpha5beta1, alpha6beta1, and heparan sulfate proteoglycans. In contrast, CCN3-induced cell migration is dependent on integrins alphavbeta3 and alpha5beta1, whereas alpha6beta1 does not play a role in this process. Although CCN3 does not contain a RGD sequence, it binds directly to immobilized integrins alphavbeta3 and alpha5beta1, with half-maximal binding occurring at 10 nm and 50 nm CCN3, respectively. Furthermore, CCN3 induces neovascularization when implanted in rat cornea, demonstrating that it is a novel angiogenic inducer. Together, these findings show that CCN3 is a ligand of integrins alphavbeta3 and alpha5beta1, acts directly upon endothelial cells to stimulate pro-angiogenic activities, and induces angiogenesis in vivo.
Sigma-Aldrich
Anti-Integrin αV Antibody, extracellular, clone AV1, azide free, culture supernatant, clone AV1, Chemicon®
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse