Merck
  • Morphological behaviour and metabolic capacity of cryopreserved human primary hepatocytes cultivated in a perfused multiwell device.
Morphological behaviour and metabolic capacity of cryopreserved human primary hepatocytes cultivated in a perfused multiwell device.
Xenobiotica; the fate of foreign compounds in biological systems (2014-07-30)
Aurelie Vivares, Sandrine Salle-Lefort, Catherine Arabeyre-Fabre, Robert Ngo, Geraldine Penarier, Michele Bremond, Patricia Moliner, Jean-François Gallas, Gerard Fabre, Sylvie Klieber
ABSTRACT
1. The quantitative prediction of the pharmacokinetic parameters of a drug from data obtained using human in vitro systems remains a significant challenge i.e. prediction of metabolic clearance in humans and estimation of the relative contribution of enzymes involved in the clearance. This has become particularly problematic for low turnover compounds. 2. Having human hepatocytes with stable cellular function over several days that adequately mimic the complexity of the physiological environment would be a major advance. Thus, we evaluated human hepatocytes, maintained in culture during 7 days in the microfluidic LiverChip™ system, in terms of morphological appearance, relative mRNA expression of phase I and II enzymes and transporters as a function of time, and metabolic capacity using probe substrates. 3. The results showed that mRNA levels of the major genes for enzymes involved in drug metabolism were well-maintained over a 7-day period of culture. Furthermore, after 4 days of culture, in the Liverchip™ device, human hepatocytes exhibited higher or similar CYPs activities compared to 1 day of culture in 2D-static conditions. 4. The functional data were supported by light/electron microscopies and immunohistochemistry showing viable tissue structure and well-differentiated human hepatocytes: presence of cell junctions, glycogen storage, and bile canaliculi.
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
2-Phenoxyethanol, ≥99%
Supelco
2-Phenoxyethanol, analytical standard
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Supelco
Linoleic acid, analytical standard
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use