Merck
  • Mesenchymal stem cells inhibit lipopolysaccharide-induced inflammatory responses of BV2 microglial cells through TSG-6.
Mesenchymal stem cells inhibit lipopolysaccharide-induced inflammatory responses of BV2 microglial cells through TSG-6.
Journal of neuroinflammation (2014-08-05)
Yi Liu, Run Zhang, Ke Yan, Fanfan Chen, Weiyi Huang, Bingke Lv, Chengmei Sun, Limin Xu, Feng Li, Xiaodan Jiang
ABSTRACT
Microglia are the primary immunocompetent cells in brain tissue and microglia-mediated inflammation is associated with the pathogenesis of various neuronal disorders. Recently, many studies have shown that mesenchymal stem cells (MSCs) display a remarkable ability to modulate inflammatory and immune responses through the release of a variety of bioactive molecules, thereby protecting the central nervous system. Previously, we reported that MSCs have the ability to modulate inflammatory responses in a traumatic brain injury model and that the potential mechanisms may be partially attributed to upregulated TNF-α stimulated gene/protein 6 (TSG-6) expression. However, whether TSG-6 exerts an anti-inflammatory effect by affecting microglia is not fully understood. In this study, we investigated the anti-inflammatory effects of MSCs and TSG-6 in an in vitro lipopolysaccharide (LPS)-induced BV2 microglial activation model. We found that MSCs and TSG-6 significantly inhibited the expression of pro-inflammatory mediators in activated microglia. However, MSC effects on microglia were attenuated when TSG-6 expression was silenced. In addition, we found that the activation of nuclear factor (NF)-κB and mitogen-activated protein kinase (MAPK) pathways in LPS-stimulated BV2 microglial cells was significantly inhibited by TSG-6. Furthermore, we found that the presence of CD44 in BV2 microglial cells was essential for MSC- and TSG-6-mediated inhibition of pro-inflammatory gene expression and of NF-κB and MAPK activation in BV2 microglial cells. The results of this study suggest that MSCs can modulate microglia activation through TSG-6 and that TSG-6 attenuates the inflammatory cascade in activated microglia. Our study indicates that novel mechanisms are responsible for the immunomodulatory effect of MSCs on microglia and that MSCs, as well as TSG-6, might be promising therapeutic agents for the treatment of neurotraumatic injuries or neuroinflammatory diseases associated with microglial activation.
Sigma-Aldrich
MISSION® esiRNA, targeting human CD44
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tnfaip6
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting human TNFAIP6
SKU
Pack Size
Availability
Price
Quantity
SAFC
L-Glutamine
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source