Merck
Topical 5-azacytidine accelerates skin wound healing in rats.
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society (2014-07-22)
Fabiana S Gomes, Gabriela F de-Souza, Lucas F Nascimento, Eva L Arantes, Rafael M Pedro, Daniele C Vitorino, Carla E Nunez, Maria H Melo Lima, Lício A Velloso, Eliana P Araújo
ABSTRACT
The development of new methods to improve skin wound healing may affect the outcomes of a number of medical conditions. Here, we evaluate the molecular and clinical effects of topical 5-azacytidine on wound healing in rats. 5-Azacytidine decreases the expression of follistatin-1, which negatively regulates activins. Activins, in turn, promote cell growth in different tissues, including the skin. Eight-week-old male Wistar rats were submitted to 8.0-mm punch-wounding in the dorsal region. After 3 days, rats were randomly assigned to receive either a control treatment or the topical application of a solution containing 5-azacytidine (10 mM) once per day. Photo documentation and sample collection were performed on days 5, 9, and 15. Overall, 5-azacytidine promoted a significant acceleration of complete wound healing (99.7% ± 0.7.0 vs. 71.2% ± 2.8 on day 15; n = 10; p < 0.01), accompanied by up to threefold reduction in follistatin expression. Histological examination of the skin revealed efficient reepithelization and cell proliferation, as evaluated by the BrdU incorporation method. 5-Azacytidine treatment also resulted in increased gene expression of transforming growth factor-beta and the keratinocyte markers involucrin and cytokeratin, as well as decreased expression of cytokines such as tumor necrosis factor-alpha and interleukin-10. Lastly, when recombinant follistatin was applied to the skin in parallel with topical 5-azacytidine, most of the beneficial effects of the drug were lost. Thus, 5-azacytidine acts, at least in part through the follistatin/activin pathway, to improve skin wound healing in rodents.
Sigma-Aldrich
Monoclonal Anti-FST antibody produced in mouse, clone 4B11, purified immunoglobulin, buffered aqueous solution