Merck
  • An improved cartilage digestion method for research and clinical applications.
An improved cartilage digestion method for research and clinical applications.
Tissue engineering. Part C, Methods (2014-09-17)
Matteo Centola, Beatrice Tonnarelli, Jeanine Hendriks, Mirella van den Doel, Sandra Feliciano, Adam Papadimitropoulos, Elia Piccinini, Jeroen Geurts, Ivan Martin, Andrea Barbero
ABSTRACT
Enzymatic isolation of chondrocytes from a cartilage biopsy is the first step to establish in vitro models of chondrogenesis or to generate cell-based grafts for cartilage repair. Such process is based on manually operated procedures and typically results in yields lower than 20% of the total available cells. In this study, we hypothesized that, as compared to conventionally used protocols, the enzymatic digestion of human articular cartilage in the presence of ascorbic acid 2-phosphate (AscA2P) or of sodium chloride (NaCl), in combination with the use of a perfusion bioreactor system, leads to a higher and more reproducible yield of cell populations with high proliferation and chondrogenic capacity. The addition of AscA2P within the enzymatic digestion medium did not significantly increase the cell yield, but resulted in a significant decrease of the intradonor variability in cell yield (-17.8% ± 10.7%, p = 0.0247) and in a significant increase of the proliferation rate of the isolated chondrocytes (+19.0% ± 1.4%, p < 0.05) with respect to the control group. The addition of NaCl during cartilage digestion did not modulate cell yield. When the cartilage digestion was further performed under direct perfusion flow, beneficial synergistic effects were achieved, with an overall increase of 34.7% ± 6.8% (p < 0.001) in the cell yield and an average decrease of 57.8% ± 11.2% (p < 0.01) in the coefficient of variation with respect to the control group. Importantly, by implementing this strategy it was possible to retrieve clonal subpopulations more efficiently capable of undergoing chondrogenesis, both in vitro and in vivo. Our findings bear relevance for the preparation of human chondrocytes for laboratory investigations, and in the perspective of efficient and streamlined manufacturing of cell/tissue grafts for articular cartilage repair.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
SAFC
HEPES
Sigma-Aldrich
Linoleic acid, ≥98%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
SAFC
HEPES
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)