- A new three-dimensional anionic cadmium(II) dicyanamide network.
A new three-dimensional anionic cadmium(II) dicyanamide network.
A new cadmium dicyanamide complex, poly[tetramethylphosphonium [μ-chlorido-di-μ-dicyanamido-κ(4)N(1):N(5)-cadmium(II)]], [(CH3)4P][Cd(NCNCN)2Cl], was synthesized by the reaction of tetramethylphosphonium chloride, cadmium nitrate tetrahydrate and sodium dicyanamide in aqueous solution. In the crystal structure, each Cd(II) atom is octahedrally coordinated by four terminal N atoms from four anionic dicyanamide (dca) ligands and by two chloride ligands. The dicyanamide ligands play two different roles in the building up of the structure; one role results in the formation of [Cd(dca)Cl]2 building blocks, while the other links the building blocks into a three-dimensional structure. The anionic framework exhibits a solvent-accessible void of 673.8 Å(3), amounting to 47.44% of the total unit-cell volume. The cavities in the network are occupied by pairs of tetramethylphosphonium cations.