Merck
  • Dual catalysis. Single-electron transmetalation in organoboron cross-coupling by photoredox/nickel dual catalysis.
Dual catalysis. Single-electron transmetalation in organoboron cross-coupling by photoredox/nickel dual catalysis.
Science (New York, N.Y.) (2014-06-07)
John C Tellis, David N Primer, Gary A Molander
ABSTRACT
The routine application of C(sp3)-hybridized nucleophiles in cross-coupling reactions remains an unsolved challenge in organic chemistry. The sluggish transmetalation rates observed for the preferred organoboron reagents in such transformations are a consequence of the two-electron mechanism underlying the standard catalytic approach. We describe a mechanistically distinct single-electron transfer-based strategy for the activation of organoboron reagents toward transmetalation that exhibits complementary reactivity patterns. Application of an iridium photoredox catalyst in tandem with a nickel catalyst effects the cross-coupling of potassium alkoxyalkyl- and benzyltrifluoroborates with an array of aryl bromides under exceptionally mild conditions (visible light, ambient temperature, no strong base). The transformation has been extended to the asymmetric and stereoconvergent cross-coupling of a secondary benzyltrifluoroborate.
Sigma-Aldrich
Methyl 3-bromobenzoate, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
[Ir{dFCF3ppy}2(bpy)]PF6
Pricing and availability is not currently available.
Sigma-Aldrich
2-(2,4-Difluorophenyl)-5-(trifluoromethyl)pyridine, ≥95%
Pricing and availability is not currently available.