Merck
Solid state characterization of oligopyridine dicarboxamide helicates.
Chemical communications (Cambridge, England) (2004-04-08)
Victor Maurizot, Gerald Linti, Ivan Huc
ABSTRACT
A heptameric amide of 2,6-diaminopyridine and 2,6-pyridinedicarboxylic acid coordinates to Cu(II) to assemble into a double helical complex with a string of six shortly spaced Cu ions.
Sigma-Aldrich
2,6-Pyridinedicarboxamide, 97%