Merck
  • Modelling the surface free energy parameters of polyurethane coats-part 1. Solvent-based coats obtained from linear polyurethane elastomers.
Modelling the surface free energy parameters of polyurethane coats-part 1. Solvent-based coats obtained from linear polyurethane elastomers.
Colloid and polymer science (2013-03-26)
Piotr Król, Jaromir B Lechowicz, Bożena Król
ABSTRACT
Polyurethane elastomers coating were synthesised by using typical diisocyanates, polyether and polyester polyols and HO-tertiary amines or diols as a chain extenders. Mole fractions of structural fragments (