Merck
  • Expression of neural cell adhesion molecule-related sialoglycoprotein in small cell lung cancer and neuroblastoma cell lines H69 and CHP-212.
Expression of neural cell adhesion molecule-related sialoglycoprotein in small cell lung cancer and neuroblastoma cell lines H69 and CHP-212.
Cancer research (1990-02-15)
C E Moolenaar, E J Muller, D J Schol, C G Figdor, E Bock, D Bitter-Suermann, R J Michalides
ABSTRACT
Monoclonal antibodies (MAbs) 123C3 and 123A8 generated against a membrane preparation of a small cell lung carcinoma (SCLC) specimen recognize not only SCLC and bronchial carcinoids but also a significant portion of non-small cell lung carcinomas (non-SCLC) of various histological types. Together with 13 other monoclonal antibodies, which show preference for SCLC, they have been ranked as SCLC cluster 1 (SC-1) Mabs. In this study we show that SC-1 MAbs are directed against a restricted number of epitopes, and that SC-1 MAbs and a polyclonal antiserum directed against the neural cell adhesion molecule (NCAM) recognize identical glycoproteins, indicating that SC-1 antigens are closely related to or identical with NCAM. Long polysialic acid units composed of alpha-(2,8)-linked N-acetylneuraminic acid units, which in mammals are found exclusively on NCAM, were present on SC-1 antigens in SCLC. This provides further evidence that SC-1 MAbs recognize NCAM. The SC-1 antigens in the SCLC cell line H69 were present in two forms, NCAM-containing alpha-(2,8)-polysialic acid units identified by antiserum 735, the NCAM-H form, and the less sialylated NCAM-L form. The NCAM-H form consisted of diffusely migrating sialoglycoproteins with a molecular weight of 200,000-250,000, which resolved after neuraminidase treatment into two proteins with molecular weights of 140,000 and 180,000. Since the NCAM-H form is expressed in the lung tumor type with a poor prognosis, our results suggest that NCAM might be implicated in the invasive behavior of these NCAM-positive lung tumors.
Sigma-Aldrich
CD56 (123C3.D5) Mouse Monoclonal Antibody, clone 123C3.D5, unconjugated, Cell Marque
Sigma-Aldrich
CD56 (MRQ-42) Rabbit Monoclonal Antibody