Merck
  • Genetic variation in the apolipoprotein H (beta2-glycoprotein I) gene affects plasma apolipoprotein H concentrations.
Genetic variation in the apolipoprotein H (beta2-glycoprotein I) gene affects plasma apolipoprotein H concentrations.
Human genetics (1999-09-10)
H Mehdi, C E Aston, D K Sanghera, R F Hamman, M I Kamboh
ABSTRACT
Apolipoprotein H (apoH, protein; APOH, gene) is a single chain glycoprotein that exists in plasma both in a free form and in combination with lipoprotein particles. ApoH has been implicated in several physiologic pathways, including lipid metabolism, coagulation, and the production of antiphospholipid antibodies. The wide range of interindividual variation in plasma apoH levels is thought to be under genetic control, but its molecular basis is unknown. APOH displays a common structural polymorphism with the occurrence of three common alleles (APOH*1, APOH*2, and APOH*3), the APOH*2 allele being the most frequent in all populations. The relationship between the APOH polymorphism and plasma apoH levels is unknown. In this study, we have determined the impact of this APOH polymorphism on apoH levels in 455 normoglycemic non-Hispanic Whites (220 men and 235 women) from the San Luis Valley, Colorado. Mean plasma apoH levels, determined by capture enzyme-linked immunosorbent assay, were 20.0 +/- 0.2 mg/dl (range: 3.4-31.2 mg/dl) with no significant difference between men and women. In women, but not in men, age had a significant effect on plasma apoH levels explaining 3.4% of its phenotypic variance. ApoH levels also correlated positively with cholesterol (P = 0.015), HDL-cholesterol (P = 0.044), and triglyceride (P = 0.037) in women, but not in men. An analysis of variance (ANOVA) of adjusted plasma apoH levels showed significant association with the APOH polymorphism in both men and women (P < 0.0001), and the APOH polymorphism accounted for 11.4% and 13.6% of the variation in apoH levels in men and women, respectively. Compared with the APOH*1 and APOH*2 alleles, the APOH*3 allele was associated with significantly lower plasma apoH levels. At the molecular level, APOH*3 can be further subdivided into two distinct forms, called APOH*3W and APOH*3B. The APOH*3W form is more common in US Whites and is the result of a missense mutation at codon 316. An ANOVA for the codon 316 polymorphism revealed that this polymorphism is a major determinant of plasma apoH variation (P < 0.0001). This study indicates that common genetic variation in the APOH gene is a significant determinant of plasma apoH levels in non-Hispanics Whites and should be useful in evaluating the role of the APOH genetic variation in various metabolic pathways in which apoH has been implicated.