Merck
  • Detecting and characterising sources of persistent organic pollutants (PAHs and PCBs) in surface sediments of an industrialized area (harbour of Trieste, northern Adriatic Sea).
Detecting and characterising sources of persistent organic pollutants (PAHs and PCBs) in surface sediments of an industrialized area (harbour of Trieste, northern Adriatic Sea).
Journal of environmental monitoring : JEM (2001-03-21)
G Adami, P Barbieri, S Piselli, S Predonzani, E Reisenhofer
ABSTRACT
A sediment sampling based on a two-dimensional mapping was performed in the harbour of Trieste (northern Adriatic Sea), considering 28 sites exposed to pollutant inputs from harbour and industrial activities. Polycyclic aromatic hydrocarbons (PAHs) and polychlorinated biphenyls (PCBs) were determined in surface sediments, because these very persistent pollutants seem to be responsible for the depletion of benthic populations observed in this area. The correlation matrix indicates that PAHs and PCBs are non-correlated, and probably have different sources. Both cluster analysis performed on the sampling sites and graphical drawing of the PAH sediment contents make it possible to locate along the shoreline a band of more polluted sediments, clustered around a site facing a steelmaking factory, to be considered as the main source point for PAHs. The evaluation of phenanthrene to anthracene (P/AN) and fluoranthene to pyrene (FL/PY) ratios permits the assessment of the pyrolytic, industrial origin of these PAHs, rejecting a second possible source of hydrocarbons (i.e., an oil-pipeline terminal, situated near the steelmaking factory). Graphical drawing of the total PCB iso-concentrations reveals a different source-point for this other category of very persistent pollutants.