MilliporeSigma
  • Role of histone H3 lysine 9 methylation in epigenetic control of heterochromatin assembly.
Role of histone H3 lysine 9 methylation in epigenetic control of heterochromatin assembly.
Science (New York, N.Y.) (2001-04-03)
J Nakayama, J C Rice, B D Strahl, C D Allis, S I Grewal
ABSTRACT
The assembly of higher order chromatin structures has been linked to the covalent modifications of histone tails. We provide in vivo evidence that lysine 9 of histone H3 (H3 Lys9) is preferentially methylated by the Clr4 protein at heterochromatin-associated regions in fission yeast. Both the conserved chromo- and SET domains of Clr4 are required for H3 Lys9 methylation in vivo. Localization of Swi6, a homolog of Drosophila HP1, to heterochomatic regions is dependent on H3 Lys9 methylation. Moreover, an H3-specific deacetylase Clr3 and a beta-propeller domain protein Rik1 are required for H3 Lys9 methylation by Clr4 and Swi6 localization. These data define a conserved pathway wherein sequential histone modifications establish a "histone code" essential for the epigenetic inheritance of heterochromatin assembly.