Merck
Alpha-linolenic acid and coronary heart disease.
Nutrition, metabolism, and cardiovascular diseases : NMCD (2004-08-28)
M de Lorgeril, P Salen
ABSTRACT
To summarize our present knowledge about vegetable omega-3 fatty acids. Alpha-linolenic acid (ALA) is one of the two essential fatty acids in humans. Epidemiological studies and dietary trials strongly suggest that this fatty acid is important in relation with the pathogenesis (and prevention) of coronary heart disease. Like other n-3 fatty acids from marine origin, it may prevent cardiac arrhythmias and sudden cardiac death. The optimal dietary intake of alpha-linolenic acid seems to be about 2 g per day or 0.6 to 1% of total energy intake. Obtaining an optimal ratio of the two essential fatty acids, linoleic and alpha-linolenic acids--ie a ratio of less than 4 to 1 in the diet--is a major issue. The main sources of alpha-linolenic acid for the European population should be canola oil (and canola-oil based margarine if available), nuts (English walnut), ground linseeds and green leafy vegetables such as purslane. Epidemiological studies and dietary trials in humans suggest that alpha-linolenic acid is a major cardio-protective nutrient.
Sigma-Aldrich
Linolenic acid, ≥99%
Sigma-Aldrich
Linolenic acid, ~70% (GC)
Supelco
Linolenic acid, analytical standard