Merck
  • Bis(2,2,2-trifluoroethyl) ether as an electrolyte co-solvent for mitigating self-discharge in lithium-sulfur batteries.
Bis(2,2,2-trifluoroethyl) ether as an electrolyte co-solvent for mitigating self-discharge in lithium-sulfur batteries.
ACS applied materials & interfaces (2014-05-17)
Mikhail L Gordin, Fang Dai, Shuru Chen, Terrence Xu, Jiangxuan Song, Duihai Tang, Nasim Azimi, Zhengcheng Zhang, Donghai Wang
ABSTRACT
Lithium-sulfur batteries suffer from severe self-discharge because of polysulfide dissolution and side reaction. In this work, a novel electrolyte containing bis(2,2,2-trifluoroethyl) ether (BTFE) was used to mitigate self-discharge of Li-S cells having both low- and high-sulfur-loading sulfur cathodes. This electrolyte meaningfully decreased self-discharge at elevated temperature, though differences in behavior of cells with high- and low-sulfur-loading were also noted. Further investigation showed that this effect likely stems from the formation of a more robust protective film on the anode surface.
Sigma-Aldrich
Bis(2,2,2-trifluoroethyl) ether, 98%