Merck
Asymmetric total synthesis of (-)-Linderol A.
The Journal of organic chemistry (2007-06-21)
Masayuki Yamashita, Navnath Dnyanoba Yadav, Takeshi Sawaki, Ikuko Takao, Ikuo Kawasaki, Yasuko Sugimoto, Akemi Miyatake, Kousuke Murai, Ayano Takahara, Ai Kurume, Shunsaku Ohta
ABSTRACT
The first asymmetric total synthesis of (-)-Linderol A, a potent inhibitor of melanin biosynthesis of cultured B-16 melanoma cells, has been achieved via two key reactions: a diastereoselective [2+2] photocycloaddition of a coumarin-3-carboxylate bearing a chiral auxiliary with 3-methyl-1-butene and a subsequent stereoconvergent transformation of the photoadducts with use of dimethylsulfoxonium methylide to afford a tetrahydrodibenzofuran derivative.