Merck
  • Towards absolute quantification of allergenic proteins in food--lysozyme in wine as a model system for metrologically traceable mass spectrometric methods and certified reference materials.
Towards absolute quantification of allergenic proteins in food--lysozyme in wine as a model system for metrologically traceable mass spectrometric methods and certified reference materials.
Journal of AOAC International (2014-03-22)
Adam Cryar, Caroline Pritchard, William Burkitt, Michael Walker, Gavin O'Connor, Duncan Thorburn Burns, Milena Quaglia
ABSTRACT
Current routine food allergen quantification methods, which are based on immunochemistry, offer high sensitivity but can suffer from issues of specificity and significant variability of results. MS approaches have been developed, but currently lack metrological traceability. A feasibility study on the application of metrologically traceable MS-based reference procedures was undertaken. A proof of concept involving proteolytic digestion and isotope dilution MS for quantification of protein allergens in a food matrix was undertaken using lysozyme in wine as a model system. A concentration of lysozyme in wine of 0.95 +/- 0.03 microg/g was calculated based on the concentrations of two peptides, confirming that this type of analysis is viable at allergenically meaningful concentrations. The challenges associated with this promising method were explored; these included peptide stability, chemical modification, enzymatic digestion, and sample cleanup. The method is suitable for the production of allergen in food certified reference materials, which together with the achieved understanding of the effects of sample preparation and of the matrix on the final results, will assist in addressing the bias of the techniques routinely used and improve measurement confidence. Confirmation of the feasibility of MS methods for absolute quantification of an allergenic protein in a food matrix with results traceable to the International System of Units is a step towards meaningful comparison of results for allergen proteins among laboratories. This approach will also underpin risk assessment and risk management of allergens in the food industry, and regulatory compliance of the use of thresholds or action levels when adopted.
Sigma-Aldrich
Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, suitable for protein sequencing, lyophilized powder
Sigma-Aldrich
Lysozyme from human neutrophils, ≥95% (SDS-PAGE), lyophilized powder, ≥100,000 units/mg protein (E1%/280)
Sigma-Aldrich
Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
Sigma-Aldrich
Trypsin from human pancreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
Sigma-Aldrich
Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin from bovine pancreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications
Sigma-Aldrich
Lysozyme human, Lysobac, recombinant, expressed in rice, ≥100,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, 10 mg/mL
Sigma-Aldrich
TrypZean® bovine, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)
Sigma-Aldrich
Lysozyme from chicken egg white, dialyzed, lyophilized, powder, ~100000 U/mg
Sigma-Aldrich
Lysozyme chloride form from chicken egg white, Grade VI, ≥35,000 units/mg protein (E1%/282)
Sigma-Aldrich
Trypsin from porcine pancreas, tablet, 1 mg tablet
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Lysozyme from chicken egg white, aseptically filled
Sigma-Aldrich
Lysozyme from chicken egg white, For use as a marker in SDS-PAGE
Supelco
Lysozyme from chicken egg white, VETRANAL®, analytical standard
Sigma-Aldrich
Trypsin from porcine pancreas, ~1500 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥90 %, ≥39,000 units/mg protein
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Lysozyme from chicken egg white, powder (crystalline), 70000-140000 U/mg