Merck
  • Hippocampal dysfunction during the chronic phase following a single exposure to cranial irradiation.
Hippocampal dysfunction during the chronic phase following a single exposure to cranial irradiation.
Experimental neurology (2014-02-05)
Yeonghoon Son, Miyoung Yang, Joong-Sun Kim, Juhwan Kim, Sung-Ho Kim, Jong-Choon Kim, Taekyun Shin, Hongbing Wang, Sung-Kee Jo, Uhee Jung, Changjong Moon
ABSTRACT
Ionizing radiation can significantly affect brain functioning in adults. The present study assessed depression-like behaviors in adult C57BL/6 mice using the tail suspension test (TST) at 30 and 90days following a single cranial exposure to γ-rays (0, 1, or 10Gy) to evaluate hippocampus-related behavioral dysfunction during the chronic phase following cranial irradiation. Additionally, hippocampal neurogenesis, inflammatory cytokines, inducible nitric oxide synthase (iNOS), cyclooxygenase-2 (COX-2), brain-derived neurotrophic factor (BDNF), and glial cell line-derived neurotrophic factor (GDNF) were analyzed. At 30 and 90days following irradiation with 10Gy, mice displayed significant depression-like behaviors. We observed a persistent decrease in the number of cells positive for doublecortin, an immunohistochemical marker for neurogenesis, in the hippocampus from 1 to 90days after irradiation with 10Gy. Changes in the mRNA expression of inflammatory cytokines, including interleukin (IL)-1β, tumor necrosis factor-α, IL-6, and interferon-γ, were not correlated with the decrease in hippocampal neurogenesis or the appearance of depression-like behavior during the chronic phase following irradiation. However, at 30 and 90days after irradiation with 10Gy, the number of microglia was significantly decreased compared with age-matched sham-irradiated controls. The reduction in the chronic phase was consistent with the significant down-regulation in the mRNA expression of iNOS, COX-2, BDNF, and GDNF in the hippocampus. Therefore, hippocampal dysfunction during the chronic phase following cranial irradiation may be associated with decreases in the neurogenesis- and synaptic plasticity-related signals, concomitant with microglial reduction in the hippocampus.
Sigma-Aldrich
Cyclooxygenase 1 from sheep, glycerol solution, ≥1500 units/mg protein
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00