Merck
  • Conditions associated with the cystic fibrosis defect promote chronic Pseudomonas aeruginosa infection.
Conditions associated with the cystic fibrosis defect promote chronic Pseudomonas aeruginosa infection.
American journal of respiratory and critical care medicine (2014-01-29)
Benjamin J Staudinger, Jocelyn Fraga Muller, Skarphéðinn Halldórsson, Blaise Boles, Angus Angermeyer, Dao Nguyen, Henry Rosen, Olafur Baldursson, Magnús Gottfreðsson, Guðmundur Hrafn Guðmundsson, Pradeep K Singh
ABSTRACT
Progress has been made in understanding how the cystic fibrosis (CF) basic defect produces lung infection susceptibility. However, it remains unclear why CF exclusively leads to chronic infections that are noninvasive and highly resistant to eradication. Although biofilm formation has been suggested as a mechanism, recent work raises questions about the role of biofilms in CF. To learn how airway conditions attributed to CF transmembrane regulator dysfunction could lead to chronic infection, and to determine if biofilm-inhibiting genetic adaptations that are common in CF isolates affect the capacity of Pseudomonas aeruginosa to develop chronic infection phenotypes. We studied P. aeruginosa isolates grown in agar and mucus gels containing sputum from patients with CF and measured their susceptibility to killing by antibiotics and host defenses. We also measured the invasive virulence of P. aeruginosa grown in sputum gels using airway epithelial cells and a murine infection model. We found that conditions likely to result from increased mucus density, hyperinflammation, and defective bacterial killing could all cause P. aeruginosa to grow in bacterial aggregates. Aggregated growth markedly increased the resistance of bacteria to killing by host defenses and antibiotics, and reduced their invasiveness. In addition, we found that biofilm-inhibiting mutations do not impede aggregate formation in gel growth environments. Our findings suggest that conditions associated with several CF pathogenesis hypotheses could cause the noninvasive and resistant infection phenotype, independently of the bacterial functions needed for biofilm formation.
Sigma-Aldrich
Elastase from human leukocytes, lyophilized powder, ≥50 units/mg protein (Bradford)