Merck
  • Late-onset Pompe disease is prevalent in unclassified limb-girdle muscular dystrophies.
Late-onset Pompe disease is prevalent in unclassified limb-girdle muscular dystrophies.
Molecular genetics and metabolism (2013-09-10)
Nicolai Preisler, Zoltan Lukacs, Lotte Vinge, Karen Lindhardt Madsen, Edith Husu, Regitze Sølling Hansen, Morten Duno, Henning Andersen, Michael Laub, John Vissing
ABSTRACT
Late-onset Pompe disease is a rare, but potentially treatable metabolic myopathy, and therefore should not be overlooked. However, it is not unusual that patients go undiagnosed for many years. We hypothesized that patients with late-onset Pompe disease may have been overlooked in a population of patients with unclassified neuromuscular disease. We used DBS (dried blood spots) to screen for Pompe disease in the two largest neuromuscular clinics and one of the main respiratory centers in Denmark. We selected patients with unclassified LGDM (limb-girdle muscular dystrophy), idiopathic elevation of creatine kinase, unexplained myopathy on muscle biopsy, unexplained restrictive respiratory insufficiency or unspecified myopathy for screening. 177 patients were found eligible for inclusion, and 103 (58.2%) patients accepted screening. Three patients with Pompe disease were identified with DBS, and subsequent genetic testing revealed known pathogenic mutations in the GAA gene. All three patients were found among 38 patients with unclassified LGMD (8%). Our findings indicate that a DBS should be considered early in the diagnostic work-up of patients with an LGMD phenotype, to rule out Pompe disease. Retrospectively, all 3 patients presented with "red flags" more compatible with Pompe disease than LGMD, including; 1) mild non-dystrophic, myopathic features on muscle biopsy, 2) creatine kinase levels below 1000, and 3) disproportionate axial and respiratory muscle involvement in comparison with limb muscle involvement.
Sigma-Aldrich
α-Glucosidase from Saccharomyces cerevisiae, Type I, lyophilized powder, ≥10 units/mg protein (using p-nitrophenyl α-D-glucoside as substrate.)
Sigma-Aldrich
α-Glucosidase from rice, Type V, ammonium sulfate suspension, 40-80 units/mg protein
Sigma-Aldrich
α-Glucosidase from Saccharomyces cerevisiae, recombinant, expressed in proprietary host, lyophilized powder, ≥100 units/mg protein
Sigma-Aldrich
α-Glucosidase from Bacillus stearothermophilus, lyophilized powder, ≥50 units/mg protein