Merck
  • Development of a novel solid-phase extraction element for thermal desorption gas chromatography analysis.
Development of a novel solid-phase extraction element for thermal desorption gas chromatography analysis.
Journal of chromatography. A (2004-05-06)
Lili Wang, Akihiko Hosaka, Chuichi Watanabe, Hajime Ohtani, Shin Tsuge
ABSTRACT
A novel solid-phase extraction element is developed for sorptive enrichment of dilute analytes from liquid samples with high extraction efficiencies due to its larger amounts of polydimethylsiloxane (PDMS) absorbent than the conventional syringe type of solid-phase microextraction (SPME). The extraction element is made of titanium (Ti) open tubular tube (30 mm x 1.2 mm i.d. x 1.6 mm o.d.) coated with a chemically bonded layer of PDMS (500 microm in thickness). The extraction element combined with thermal desorption-gas chromatography-mass spectrometry using a pyrolysis-gas chromatography-mass spectrometry system was used to extract and analyze a typical herbicide, bethrodine in water samples over a concentration range from 2.5 to 2.5 x 10(4) ng/l. Thus obtained calibration curve showed good linearity for the tested whole concentration range with regression coefficient of 0.992. Detection limit of 0.5 ng/l level was achieved and the reproducibility of the measurements for bethrodine at 10 ng/l level was found to be fairly good with relative standard deviation below 7.5%.
Product Description
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 23 ga, for use with autosampler