Merck
In vivo depigmentation by hydroxybenzene derivatives.
Melanoma research (1993-12-01)
J M Menter, A A Etemadi, W Chapman, T D Hollins, I Willis
ABSTRACT
Certain mono- and dihydroxybenzene derivatives are selectively cytotoxic for melanocytes in vivo, and can cause depigmentation of skin and hair. We produced selective melanocytotoxicity/hair depigmentation in C57Bl mice by injection of 0.032-1.0% p-t-butylcatechol (tBC) or p-hydroxyanisole (MMEH) in physiological saline. No depigmentation occurred on injection of 3,4-dihydroxyphenylalanine (DOPA) or 3,4-dihydroxyphenylacetic acid (DOPAC). Light- and electron-microscopic examination of biopsy specimens taken from depigmented areas indicates selective melanocyte damage as early as 2 h post-injection. Melanocytes from anagen hair are most susceptible to depigmentation. All four compounds are substrates for tyrosinase, but only tBC and MMEH generate their respective isolable 1,2-benzoquinones, tBCQ and MMEHQ. These caused depigmentation in C57Bl mice to a comparable degree to the parent compounds. DOPA- and DOPAC-quinones (DOPAQ and DOPACQ) are not spectroscopically detectable in solution, suggesting extremely low steady-state levels of these compounds. The net observed rate of reaction of the respective 1,2-quinone with 300 microM bovine serum albumin (BSA) in vitro varies widely, with tBCQ > MMEHQ = DOPACQ > DOPAQ. The results are consistent with a mechanism involving attack of -SH on melanosomal proteins and/or enzymes by tyrosinase-generated 1,2-quinones. This mechanism evidently differs from that involved in in vitro hydroxybenzene melanocytotoxicity of melanoma cells, in which active oxygen intermediates generated by hydroxybenzene autoxidation play a significant role. The most reliable prognosticator of in vivo depigmentation appears to be the ability of the depigmenter to form a spectroscopically stable 1,2-quinone which is capable of reacting with protein -SH.
Product Description
USP
2-tert-Butyl-4-hydroxyanisole, United States Pharmacopeia (USP) Reference Standard