Merck
  • The advantages of topical combination therapy in the treatment of inflammatory dermatomycoses.
The advantages of topical combination therapy in the treatment of inflammatory dermatomycoses.
Mycoses (2008-09-12)
Blanka Havlickova, Markus Friedrich
ABSTRACT
Dermatomycoses are contagious superficial fungal infections, which are highly prevalent in developed and developing countries. Caused by a range of Epidermophyton, Microsporum and Trichophyton species, dermatomycoses manifest on glabrous skin as 'ringworm', an annular scaly lesion with a variable inflammatory component. Itch is the chief subjective symptom, particularly in tinea cruris. Unless lesions are extensive or resistant to local therapy, dermatomycoses of glabrous skin are treated with topical antifungal agents, such as imidazoles and allylamines. Studies show, however, that the addition of a topical corticosteroid to imidazole therapy increases the bioavailability and prolongs the activity of the antimycotic, while rapidly reducing inflammatory symptoms. Travocort is a combination of 1% isoconazole nitrate (ISN), a broad-spectrum imidazole with established antimicrobial activity and antimycotic efficacy, and 0.1% diflucortolone valerate (DFV), a potent topical corticosteroid with low systemic absorption and therefore a low risk of systemic glucocorticoid side-effects. In randomised, double-blind controlled clinical trials, Travocort therapy showed a more rapid onset of action, faster relief of itch and other inflammatory symptoms, improved overall therapeutic benefits and better mycological cure rate during the first 2 weeks of treatment compared with ISN monotherapy. Travocort is well tolerated and, because of prolonged ISN retention in the skin, provides antifungal protection against reinfection for some weeks after therapy.
Product Description
Isoconazole nitrate, European Pharmacopoeia (EP) Reference Standard
Isoconazole, European Pharmacopoeia (EP) Reference Standard