Merck
  • Sequestration, metabolism and partial synthesis of tertiary pyrrolizidine alkaloids by the neotropical leaf-beetle Platyphora boucardi.
Sequestration, metabolism and partial synthesis of tertiary pyrrolizidine alkaloids by the neotropical leaf-beetle Platyphora boucardi.
Insect biochemistry and molecular biology (2001-08-25)
T Hartmann, C Theuring, L Witte, J M Pasteels
ABSTRACT
Platyphora boucardi leaf-beetles sequester tertiary pyrrolizidine alkaloids of the lycopsamine type acquired from their host-plant Prestonia portobellensis (Apocynaceae) and synthesize their own alkaloids from exogenous retronecine and aliphatic 2-hydroxy acids. Tracer studies with [14C]rinderine and its N-oxide revealed that P. boucardi sequesters both alkaloidal forms with the same efficiency, but accumulates exclusively tertiary alkaloids. There is no substantial alkaloid accumulation in the body outside the defensive glands. Feeding studies with [2H][14C]rinderine confirmed that P. boucardi specifically epimerizes rinderine to its stereoisomers intermedine and lycopsamine. Feeding studies with [2H][14C]retronecine proved the ability of P. boucardi to synthesize O7- and O9-(2-hydroxyisovaleryl)-retronecine and O7-lactyl-O9-(2-hydroxyisovaleryl)-retronecine. Both, alkaloids of the lycopsamine type and self-synthesized retronecine esters accumulate in the defensive secretions at concentrations up to 38 mM and 33 mM, respectively. The different biochemical strategies to maintain pro-toxic pyrrolizidine alkaloids and to prevent self-poisoning, developed by specialized insects, are compared. There are two major findings: (1) the chemical defense mediated by plant acquired pyrrolizidine alkaloids in the taxonomically related palaearctic Oreina and neotropical Platyphora leaf beetles have been evolved independently, since the biochemical mechanisms of storing and maintaining the alkaloids is completely different in the two genera; (2) unexpected parallels exist between taxonomically unrelated Coleoptera and Lepidoptera in their ability to synthesize the same retronecine esters and to catalyze the same site-specific epimerizations of the lycopsamine stereoisomers.