Merck
  • Adenosine deaminase inhibitors. Synthesis and biological evaluation of C1' and nor-C1' derivatives of (+)-erythro-9-(2(S)-hydroxy-3(R)-nonyl)adenine.
Adenosine deaminase inhibitors. Synthesis and biological evaluation of C1' and nor-C1' derivatives of (+)-erythro-9-(2(S)-hydroxy-3(R)-nonyl)adenine.
Journal of medicinal chemistry (1992-10-30)
G C Harriman, A F Poirot, E Abushanab, R M Midgett, J D Stoeckler
ABSTRACT
The synthesis of various chiral derivatives of (+)-erythro-9-(2-hydroxy-3-nonyl)adenine, (+)-EHNA, from (2S,3R)-3-amino-1,2-O-isopropylidene-1,2-nonanediol by condensation with 5-amino-4,6-dichloropyrimidine is described. The compounds synthesized were C1'- and nor-C1'-(+)-EHNA derivatives. When tested with calf spleen ADA, C1'-OH- and nor-C1'-(+)-EHNA had comparable inhibitory activity that was 1 order of magnitude lower than that of (+)-EHNA. Potency was reduced further in nor-C1' derivatives.
Sigma-Aldrich
5-Amino-4,6-dichloropyrimidine, 97%