Merck
  • Amylase α-1A (AMY1A): a novel immunohistochemical marker to differentiate chromophobe renal cell carcinoma from benign oncocytoma.
Amylase α-1A (AMY1A): a novel immunohistochemical marker to differentiate chromophobe renal cell carcinoma from benign oncocytoma.
The American journal of surgical pathology (2013-11-15)
Sarika Jain, Somak Roy, Milon Amin, Marie Acquafondata, Ming Yin, William Laframboise, Sheldon Bastacky, Liron Pantanowitz, Rajiv Dhir, Anil Parwani
ABSTRACT
Chromophobe renal cell carcinoma (ChRCC) and oncocytoma present with a perplexing overlap of morphologic and immunohistochemical features. ChRCC have deletions in the 1p21.1 region including the amylase α-1A gene (AMY1A). No such deletions are found in oncocytoma. Instead, oncocytomas shared other deletions on chromosome 1: 1p31.3, 1q25.2, and 1q44. We performed AMY1A immunostaining on 75 oncocytomas (57 tissue microarray [TMA] cores, 18 whole slides) and 54 ChRCCs (20 TMA cores, 34 whole slides). Staining was assessed using the H-score method. The intensity was graded as follows: no staining=0, weak=1, moderate=2, and strong=3. The AMY1A immunostain preferentially stained the distal tubules and collecting ducts of normal kidney. All oncocytomas (100%) expressed AMY1A with an H-score that varied from 100 to 300 (mean 205). Mild to moderate heterogeneity in staining intensity was noted within a given oncocytoma. For oncocytomas, 87% (65/75) cases had H-scores of at least 120 with a mean score of 221. Notably, the 13% (10/75) of oncocytoma cases that had an H-score of 100 were derived from the TMA. A total of 87% (47/54) of the ChRCC cases were negative for the AMY1A immunostain. Of the ChRCC cases, 4% (2/54) showed very weak cytoplasmic staining (H-score of 70 each), which was less than the lowest H-score of oncocytoma cases. All 5 cases of ChRCC, which showed an H-score of 100 or more, were referred to as eosinophilic variants of ChRCC. Three of these 5 cases showed a very nondescript, diffuse staining of the cytoplasm. Two of these 5 cases showed an H-score of 130. We think that as the staining pattern of these 2 cases is similar to that of oncocytoma, they should be put in a category of renal oncocytic neoplasms favoring oncocytoma. This result shows that AMY1A staining could be very helpful in further classifying even a subset of the eosinophilic variants of ChRCC. The difference between ChRCC and oncocytoma was statistically significant (χ test, P<0.0001). All cases of clear cell RCC and papillary RCC were negative for AMY1A expression. Overall, sensitivity and specificity of AMY1A staining for oncocytoma was 100% (95% confidence interval, 0.95-1.00) and 96.75% (95% confidence interval, 0.93-0.99), respectively. Similarly, the sensitivity and specificity for distinguishing oncocytoma from ChRCC was 100% (95% confidence interval, 0.95-1.00) and 90.74% (95% confidence interval, 0.80-0.97), respectively. These data show that the novel marker AMY1A can be of great diagnostic utility when trying to differentiate ChRCC (classic and eosinophilic variant) and oncocytoma.
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg