Merck
  • Metoclopramide in pregnancy and risk of major congenital malformations and fetal death.
Metoclopramide in pregnancy and risk of major congenital malformations and fetal death.
JAMA (2013-10-17)
Björn Pasternak, Henrik Svanström, Ditte Mølgaard-Nielsen, Mads Melbye, Anders Hviid
ABSTRACT
Metoclopramide, a drug frequently used for nausea and vomiting in pregnancy, is thought to be safe, but information on the risk of specific malformations and fetal death is lacking. To investigate the safety of metoclopramide use in pregnancy. Register-based cohort study in Denmark, 1997-2011. From a cohort of 1,222,503 pregnancies, metoclopramide-exposed and unexposed women were matched (1:4 ratio) on the basis of age, calendar year, and propensity scores. Primary outcomes were major congenital malformations overall, 20 individual malformation categories (selected according to power criteria), spontaneous abortion, and stillbirth. In matched analyses, logistic regression was used to estimate prevalence odds ratios of malformations and Cox regression to estimate hazard ratios (HRs) of spontaneous abortion. Among 28,486 women exposed to metoclopramide in the first trimester, 721 had an infant with a major congenital malformation (25.3 [95% CI, 23.5-27.1] cases per 1000 births), compared with 3024 among 113,698 unexposed women (26.6 [95% CI, 25.7-27.5] per 1000 births). There were no significant associations between metoclopramide use and malformations overall (prevalence odds ratio, 0.93 [95% CI, 0.86-1.02]) or any of the 20 individual malformation categories, eg, neural tube defects, transposition of great vessels, ventricular septal defect, atrial septal defect, tetralogy of Fallot, coarctation of the aorta, cleft lip, cleft palate, anorectal atresia/stenosis, and limb reduction (upper limit of 95% CI below 2.0 for 17 of 20 categories). Metoclopramide was not associated with increased risk of spontaneous abortion (757 cases [20.0 {95% CI, 18.5-21.4} per 1000] among 37,946 metoclopramide-exposed women and 9414 cases [62.1 {95% CI, 60.9-63.3} per 1000] among 151,661 unexposed women; HR, 0.35 [95% CI, 0.33-0.38]) and stillbirth (142 cases [3.5 {95% CI, 2.9-4.1} per 1000] among 40,306 metoclopramide-exposed women and 634 cases [3.9 {95% CI, 3.6-4.2} per 1000] among 161,098 unexposed women; HR, 0.90 [95% CI, 0.74-1.08]). Metoclopramide use in pregnancy was not associated with increased risk of major congenital malformations overall, any of the 20 individual malformation categories assessed, spontaneous abortion, or stillbirth. These safety data may help inform decision making when treatment with metoclopramide is considered in pregnancy.
Product Description
Supelco
Metoclopramide, VETRANAL®, analytical standard
USP
Metoclopramide hydrochloride, United States Pharmacopeia (USP) Reference Standard