Merck
  • A novel array of chemiluminescence sensors for sensitive, rapid and high-throughput detection of explosive triacetone triperoxide at the scene.
A novel array of chemiluminescence sensors for sensitive, rapid and high-throughput detection of explosive triacetone triperoxide at the scene.
Biosensors & bioelectronics (2013-04-24)
Xiaohua Li, Zhujun Zhang, Liang Tao
ABSTRACT
Triacetone triperoxide (TATP) is relatively easy to make and has been used in various terrorist acts. Early but easy detection of TATP is highly desired. We designed a new type sensor array for H2O2. The unique CL sensor array was based on CeO2 nanoparticles' membranes, which have an excellent catalytic effect on the luminol-H2O2 CL reaction in alkaline medium. It exhibits a linear range for the detection of H2O2 from 1.0×10(-8) to 5.0×10(-5)M (R(2)=0.9991) with a 1s response time. The detection limit is 1.0×10(-9)M. Notably, the present approach allows the design of CL sensor array assays in a more simple, time-saving, long-lifetime, high-throughput, and economical approach when compared with conventional CL sensor. It is conceptually different from conventional CL sensor assays. The novel sensor array has been successfully applied for the detection of TATP at the scene.
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <50 nm particle size (BET), 99.95% trace rare earth metals basis
Sigma-Aldrich
Cerium(IV) oxide, powder, <5 μm, 99.9% trace metals basis
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Cerium(IV) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <25 nm particle size (BET)
Sigma-Aldrich
Cerium(IV) oxide, dispersion, 20 wt. % colloidal dispersion in 2.5% acetic acid, 30-50 nm avg. part. size
Sigma-Aldrich
Cerium(IV) oxide, ≥99.0%
Sigma-Aldrich
Cerium, chips
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Cerium(IV) oxide, dispersion, nanoparticles, <25 nm particle size, 10 wt. % in H2O