Merck
  • Intranasal delivery of cell-penetrating anti-NF-κB peptides (Tat-NBD) alleviates infection-sensitized hypoxic-ischemic brain injury.
Intranasal delivery of cell-penetrating anti-NF-κB peptides (Tat-NBD) alleviates infection-sensitized hypoxic-ischemic brain injury.
Experimental neurology (2013-01-29)
Dianer Yang, Yu-Yo Sun, Xiaoyi Lin, Jessica M Baumann, R Scott Dunn, Diana M Lindquist, Chia-Yi Kuan
ABSTRACT
Perinatal infection aggravates neonatal hypoxic-ischemic (HI) brain injury and may interfere with therapeutic hypothermia. While the NF-κB signaling pathway has been implicated in microglia activation in infection-sensitized HI, the current therapeutic strategies rely on systemic intervention, which could impair neonatal immunity and increase the risk of severe infection. To devise a brain-targeted anti-NF-κB strategy, we examined the effects of intranasal delivery of tat-NBD peptides in two animal models of neonatal infection-sensitized HI. Kinetic experiments showed that tat-NBD peptides entered the olfactory bulbs rapidly (10-30 min) and peaked in the cerebral cortex around 60 min after intranasal application in P7 rats. Further, intranasal delivery of 1.4 mg/kg tat-NBD, which is only 7% of the intravenous dose in past studies, markedly attenuated NF-κB signaling, microglia activation, and brain damage triggered by HI with 4 or 72 h pre-exposure to the bacterial endotoxin lipopolysaccharide (LPS). In contrast, intranasal delivery of mutant tat-NBD peptides or systemic application of minocycline failed to block LPS-sensitized HI injury. Yet, intranasal delivery of up to 5.6 mg/kg tat-NBD peptides immediately after pure-HI insult showed little protection, likely due to its rapid clearance from the brain and inability to inhibit parenchymal plasminogen activators. Together, these results suggest a novel therapy of infection-sensitized HI brain injury in newborns.
Product Description
Millipore
2,3,5-Triphenyl-tetrazolium chloride solution, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC
Pricing and availability is not currently available.
Sigma-Aldrich
2,3,5-Triphenyltetrazolium chloride, ≥98.0% (HPLC)
Pricing and availability is not currently available.