Merck
  • TempliPhi: A sequencing template preparation procedure that eliminates overnight cultures and DNA purification.
TempliPhi: A sequencing template preparation procedure that eliminates overnight cultures and DNA purification.
Journal of biomolecular techniques : JBT (2003-12-17)
Michael J Reagin, Theresa L Giesler, Alia L Merla, Jeanine M Resetar-Gerke, Kinga M Kapolka, J Anthony Mamone
ABSTRACT
Preparing plasmid templates for DNA sequencing is the most time-consuming step in the sequencing process. Current template preparation methods rely on a labor-intensive, multistep procedure that takes up to 24 h and produces templates of varying quality and quantity. The TempliPhi DNA Sequencing Template Amplification Kit eliminates the requirement for extended bacterial growth prior to sequencing and saves laboratory personnel hands-on time by eliminating the centrifugation and transfer steps currently required by older preparatory methods. In addition, costly purification filters and columns are not necessary, as amplified product can be added directly to a sequencing reaction. Starting material can be any circular template from a colony, culture, glycerol stock, or plaque. Based on rolling circle amplification and employing bacteriophage Phi29 DNA polymerase, the method can produce 3-5 microg of template directly from a single bacterial colony in as little as 4 h. Implementation of these procedures in a laboratory or core sequencing facility can decrease cost on tips, plates, and other plasticware, while at the same time increase throughput.
Product Description
TempliPhi 500 Amplification Kit, Cytiva 25-6400-50
TempliPhi - 2000 Reaction Kit, Cytiva 28-9642-86
TempliPhi 100 Amplification Kit, Cytiva 25-6400-10