Merck
  • Involvement of carnitine acyltransferases in peroxisomal fatty acid metabolism by the yeast Pichia guilliermondii.
Involvement of carnitine acyltransferases in peroxisomal fatty acid metabolism by the yeast Pichia guilliermondii.
Applied and environmental microbiology (1996-10-01)
Y Pagot, J M Belin
ABSTRACT
This article provides information about peroxisomal fatty acid metabolism in the yeast Pichia guilliermondii. The existence of inducible mitochondrial carnitine palmitoyltransferase and peroxisomal carnitine octanoyl-transferase activities was demonstrated after culture of this yeast in a medium containing methyl oleate. The subcellular sites and induction patterns were studied. The inhibition of carnitine octanoyl- and palmitoyl-transferases by chlorpromazine to a large extent prevented the otherwise observed metabolism-dependent inactivation of thiolase by 2-bromofatty acids in vivo. We concluded that the metabolism of long- and medium-chain fatty acids in the peroxisome of this yeast involved carnitine intermediates.