- Novel pyrimidines as acid pump antagonists (APAs).
Novel pyrimidines as acid pump antagonists (APAs).
Bioorganic & medicinal chemistry letters (2010-09-03)
Young Ae Yoon, Chan Sun Park, Myung Hun Cha, Hyunho Choi, Jae Young Sim, Jae Gyu Kim
PMID20810280
ABSTRACT
A series of pyrimidine derivatives as acid pump antagonists (APAs) was synthesized and the inhibitory activities against H(+)/K(+) ATPase isolated from hog gastric mucosa were determined. After elaborating on substituents at C2 and C4 position of the pyrimidine scaffold, we have observed that the compound 7h is a potent APA with H(+)/K(+) ATPase, IC(50) = 52 nM.