Merck
  • Efficient synthesis and characterization of novel bibenzimidazole oligomers and polymers as potential conjugated chelating ligands.
Efficient synthesis and characterization of novel bibenzimidazole oligomers and polymers as potential conjugated chelating ligands.
The Journal of organic chemistry (2005-11-05)
Jun Yin, Ronald L Elsenbaumer
ABSTRACT
[Structure: see text]. A simple and mild condensation route for the synthesis of novel bibenzimidazole oligomers and polymers is reported here using methyl 2,2,2-trichloroacetimidate as a key starting material. The dimer, trimer, tetramer, and polymers of bibenzimidazole were synthesized as a new series of potential conjugated chelating ligands for metallopolymer studies. The polymers show a maximum absorption at around 400 nm. The optical band gap of the polymer was estimated to be 2.68 eV.
Sigma-Aldrich
Methyl 2,2,2-trichloroacetimidate, 98%