Merck
  • Determination of reduced and oxidized glutathione in erythrocytes by high-performance liquid chromatography with ultraviolet absorbance detection.
Determination of reduced and oxidized glutathione in erythrocytes by high-performance liquid chromatography with ultraviolet absorbance detection.
Journal of chromatography. B, Biomedical applications (1996-04-12)
T Yoshida
ABSTRACT
A method is described for simultaneous quantitation of reduced (GSH) and oxidized (GSSG) glutathione in erythrocytes by HPLC. They were determined by standard addition method. Blood samples were collected in tubes containing 1,10-phenanthroline. The separated erythrocytes were hemolyzed with water containing standard. After deproteinization, GSH and GSSG were converted to N-(2,4-dinitrophenyl) derivatives and analyzed by HPLC with UV detection. The coefficients of variation of GSH and GSSG on replicate assays were 6% and 8%, respectively. The stabilities of GSH and GSSG and of the derivatives were also examined. The present method appears to be satisfactory for determination of these physiological concentrations in erythrocytes.
Sigma-Aldrich
L-Glutathione oxidized, BioXtra, ≥98%
Sigma-Aldrich
L-Glutathione oxidized, ≥98% (HPLC)
Sigma-Aldrich
L-Glutathione oxidized, lyophilized powder