Merck
  • Inhibition of Listeria monocytogenes and Salmonella enteriditis by combinations of plant oils and derivatives of benzoic acid: the development of synergistic antimicrobial combinations.
Inhibition of Listeria monocytogenes and Salmonella enteriditis by combinations of plant oils and derivatives of benzoic acid: the development of synergistic antimicrobial combinations.
International journal of antimicrobial agents (1997-01-01)
L Fyfe, F Armstrong, J Stewart
ABSTRACT
This study describes inhibitory properties of combinations of oil of fennel, oil of anise or oil of basil with either benzoic acid or methyl-paraben against Listeria monocytogenes and Salmonella enteriditis. Micro-organisms were cultured at 37 degrees C in broth and viable counts measured over a 48-h period. S. enteriditis was particularly sensitive to inhibition by a combination of oil of anise, fennel or basil with methyl-paraben where there was < 10 CFU/ml after 1 h. L. monocytogenes was less sensitive to inhibition by each combination however there was a significant reduction in growth of 4-8 log by combinations of all oils and methyl-paraben at 8, 24 and 48 h. Synergistic inhibition by one or more combinations was evident against each micro-organism.