Merck
  • Inclusion of the main pheromone component of Dacus oleae, 1,7-dioxaspiro[5,5]undecane, in beta-cyclodextrin.
Inclusion of the main pheromone component of Dacus oleae, 1,7-dioxaspiro[5,5]undecane, in beta-cyclodextrin.
Acta crystallographica. Section C, Crystal structure communications (1996-11-15)
N Rysanek, G Le Bas, F Villain, G Tsoucaris
ABSTRACT
The complex beta-cyclodextrin-1,7-dioxaspiro[5,5]undecane nonahydrate, C42H70O35.C9H16O2.9H2O, belongs to the class of beta-cyclodextrin dimeric-type complexes. The racemic guest molecule is present in a disordered position. Both enantiomers are located in two different regions inside the channel formed by the host dimers.
Sigma-Aldrich
1,7-Dioxaspiro[5.5]undecane, ≥97%