Merck
  • Structural elucidation of two novel ergot alkaloid impurities in alpha-ergokryptine and bromokryptine.
Structural elucidation of two novel ergot alkaloid impurities in alpha-ergokryptine and bromokryptine.
Acta pharmaceutica Hungarica (1994-05-01)
C Szántay, M Bihari, J Brlik, A Csehi, A Kassai, A Aranyi
ABSTRACT
Two novel natural derivatives (2 and 3) of the ergot alkaloid alpha-ergokryptine (1), as well as their synthetically brominated analogues (5 and 6) were isolated and identified by NMR and MS methods. Compounds 2 and 5 contain what appears to be a so far unknown natural amino acid building block. Complete 1H and 13C NMR assignments are given for compounds 1-6.