- Leukotriene C synthase in mouse mastocytoma cells. An enzyme distinct from cytosolic and microsomal glutathione transferases.
Leukotriene C synthase in mouse mastocytoma cells. An enzyme distinct from cytosolic and microsomal glutathione transferases.
Leukotriene C4 synthesis was studied in preparations from mouse mastocytoma cells. Enzymic conjugation of leukotriene A4 with glutathione was catalysed by both the cytosol and the microsomal fraction. The specific activity of the microsomal fraction (7.8 nmol/min per mg of protein) was 17 times that of the cytosol fraction. The cytosol fraction of the mastocytoma cells contained two glutathione transferases, which were purified to homogeneity and characterized. A microsomal glutathione transferase was purified from mouse liver; this enzyme was shown by immunoblot analysis to be present in the mastocytoma microsomal fraction at a concentration one-tenth or less of that in the liver microsomal fraction. Both the cytosolic and the microsomal glutathione transferases in the mastocytoma cells were identified with enzymes previously characterized, by determining specific activities with various substrates, sensitivities to inhibitors, reactions with antibodies, and physical properties. The purified microsomal glutathione transferase from liver was inactive with leukotriene A4 or its methyl ester as substrate. The cytosolic enzymes displayed activity with leukotriene A4, but their specific activities and intracellular concentrations were too low to account for the leukotriene C4 formation in the mastocytoma cells. The microsomal fraction of the cells contained an enzyme distinguishable by various criteria from the previously studied glutathione transferases. This membrane-bound enzyme, leukotriene C synthase (leukotriene A4:glutathione S-leukotrienyltransferase), appears to carry the main responsibility for the biosynthesis of leukotriene C4.