Merck
  • [Hypotensive, antianginal action and pharmacokinetics of glyceryl-1-nitrate in rats and dogs].
[Hypotensive, antianginal action and pharmacokinetics of glyceryl-1-nitrate in rats and dogs].
Arzneimittel-Forschung (1986-11-01)
M Leitold, H Laufen, R A Yeates
ABSTRACT
The antianginal and hypotensive activity and the pharmacokinetic properties of glyceryl 1-nitrate (G-1-N) were examined in the rat and in the dog. The level and duration of antianginal and hypotensive activity were the same after single or repeated oral dosage of G-1-N to the anaesthetized rat. The haemodynamic activity of intravenously administered G-1-N in the anaesthetized and thoracotomized dog was dose-dependent. A bolus injection of G-1-N or glyceryl trinitrate (GTN, Nitro Mack) antagonized the cardiovascular activity of intravenously injected dihydroergotamine (DHE) in the anaesthetized and thoracotomized dog. The bioavailability of G-1-N in the rat and in the dog is practically 100%. After intravenous or oral administration to the rat the concentrations of G-1-N in the walls of the vena cava caudalis were markedly higher and in the aorta abdominalis somewhat higher than in the blood or in the other organs examined. G-1-N was eliminated more slowly from the walls of the animals veins than from the walls of the aorta abdominalis or from the blood.
Product Description
Supelco
1-Mononitroglycerin solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®